Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515340.1 | XP_003515388.1 | 4 - 5 | 698 | 63 | |
XM_007607553.1 | XP_007605743.1 | 4 - 5 | 698 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |