Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652779.1 | XP_007650969.1 | 2 - 3 | 248 | 98 | |
GenBank mRNA | JP047327.1 | - | 428 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |