Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641811.1 | XP_007640001.1 | 2 - 3 | 119 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Probe spans exons |