Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503173.1 | XP_003503221.1 | 1 - 2 | 84 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |