Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505458.2 | XP_003505506.2 | 5 - 6 | 715 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |