Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506801.1 | XP_003506849.1 | 2 - 3 | 699 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Probe spans exons |