Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Errfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513303.2 | XP_003513351.1 | 1 - 2 | 388 | 97 | |
GenBank mRNA | JI875159.1 | - | 1 - 2 | 215 | 97 | |
JP046867.1 | - | 1 - 2 | 215 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |