Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514754.2 | XP_003514802.1 | 5 - 6 | 1235 | 88 | |
XM_007654811.1 | XP_007653001.1 | 5 - 6 | 1040 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |