Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646682.1 | XP_007644872.1 | 3 - 4 | 758 | 64 | |
GenBank mRNA | JP038954.1 | - | 2 - 3 | 288 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |