Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007637258.1 | XP_007635448.1 | 1 - 2 | 306 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |