Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652014.1 | XP_007650204.1 | 2 - 3 | 171 | 114 | |
GenBank mRNA | JI877238.1 | - | 1 - 2 | 112 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |