Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511823.1 | XP_003511871.1 | 12 - 13 | 1121 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |