Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513852.2 | XP_003513900.1 | 2 - 3 | 395 | 67 | |
XM_007654065.1 | XP_007652255.1 | 2 - 3 | 209 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |