Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514533.2 | XP_003514581.1 | 1 - 2 | 213 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |