Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646941.1 | XP_007645131.1 | 1 - 2 | 78 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |