Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504686.2 | XP_003504734.1 | 6 - 7 | 753 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |