Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496150.2 | XP_003496198.1 | 1 - 2 | 280 | 108 | |
XM_007647313.1 | XP_007645503.1 | 1 - 2 | 280 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |