Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497291.2 | XP_003497339.1 | 1 | 423 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Ion Channel Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |