Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503117.1 | XP_003503165.1 | 2 - 3 | 930 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Probe spans exons |