Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510396.2 | XP_003510444.1 | 6 - 7 | 827 | 87 | |
XM_007651026.1 | XP_007649216.1 | 6 - 7 | 867 | 87 | ||
GenBank mRNA | JI873405.1 | - | 6 - 7 | 817 | 87 | |
JP064031.1 | - | 6 - 7 | 817 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Probe spans exons |