Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647825.1 | XP_007646015.1 | 9 - 10 | 3882 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |