Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503637.1 | XP_003503685.1 | 1 - 2 | 180 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |