Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648922.1 | XP_007647112.1 | 2 - 3 | 408 | 89 | |
GenBank mRNA | JI882986.1 | - | 2 - 3 | 408 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |