Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497688.2 | XP_003497736.1 | 5 - 6 | 1000 | 98 | |
XM_007615896.1 | XP_007614086.1 | 6 - 7 | 1091 | 98 | ||
GenBank mRNA | JI882145.1 | - | 5 - 6 | 1002 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |