Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510832.2 | XP_003510880.1 | 3 - 4 | 365 | 81 | |
GenBank mRNA | JI893460.1 | - | 3 - 4 | 327 | 81 | |
JP038674.1 | - | 3 - 4 | 327 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |