Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642311.1 | XP_007640501.1 | 3 - 4 | 432 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |