Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503318.1 | XP_003503366.1 | 1 - 2 | 386 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |