Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650055.1 | XP_007648245.1 | 2 - 3 | 278 | 99 | |
GenBank mRNA | JI875058.1 | - | 2 - 3 | 278 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Detect Genomic DNA Probe spans exons |