Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495323.2 | XP_003495371.1 | 3 - 4 | 1865 | 83 | |
XM_007637971.1 | XP_007636161.1 | 4 - 5 | 1951 | 83 | ||
XM_007638031.1 | XP_007636221.1 | 3 - 4 | 1612 | 83 | ||
GenBank mRNA | JI869930.1 | - | 1 - 2 | 776 | 83 |