Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495323.2 | XP_003495371.1 | 7 - 8 | 2516 | 76 | |
XM_007637971.1 | XP_007636161.1 | 8 - 9 | 2602 | 76 | ||
XM_007638031.1 | XP_007636221.1 | 7 - 8 | 2263 | 76 | ||
GenBank mRNA | JI869930.1 | - | 5 - 6 | 1428 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |