Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfnb31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503466.2 | XP_003503514.1 | 6 - 7 | 744 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |