Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510181.2 | XP_003510229.1 | 1 - 2 | 38 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |