Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007621739.1 | XP_007619929.1 | 1 - 2 | 382 | 80 | |
XM_007621745.1 | XP_007619935.1 | 1 - 2 | 382 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |