Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007621753.1 | XP_007619943.1 | 1 - 2 | 336 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |