Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510698.2 | XP_003510746.1 | 2 - 3 | 195 | 95 | |
GenBank mRNA | JI875411.1 | - | 2 - 3 | 183 | 95 | |
JP042038.1 | - | 2 - 3 | 183 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |