Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499003.1 | XP_003499051.1 | 1 - 2 | 105 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |