Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508154.2 | XP_003508202.1 | 4 - 5 | 351 | 76 | |
GenBank mRNA | JI881738.1 | - | 4 - 5 | 356 | 76 | |
JP062017.1 | - | 4 - 5 | 356 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |