Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508615.1 | XP_003508663.1 | 2 - 3 | 401 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |