Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642784.1 | XP_007640974.1 | 3 - 4 | 405 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |