Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648251.1 | XP_007646441.1 | 8 - 9 | 1852 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |