Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497613.2 | XP_003497661.1 | 3 | 249 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Both primers and probe map within a single exon |