Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650852.1 | XP_007649042.1 | 8 - 9 | 1119 | 75 | |
XM_007650853.1 | XP_007649043.1 | 8 - 9 | 1101 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |