Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504701.2 | XP_003504749.1 | 5 - 6 | 1910 | 78 | |
XM_007645214.1 | XP_007643404.1 | 9 - 10 | 4661 | 78 | ||
XM_007645215.1 | XP_007643405.1 | 9 - 10 | 4453 | 78 | ||
XM_007645216.1 | XP_007643406.1 | 9 - 10 | 4440 | 78 | ||
XM_007645217.1 | XP_007643407.1 | 9 - 10 | 4574 | 78 | ||
XM_007645218.1 | XP_007643408.1 | 8 - 9 | 4490 | 78 | ||
XM_007645219.1 | XP_007643409.1 | 8 - 9 | 4387 | 78 | ||
XM_007645220.1 | XP_007643410.1 | 8 - 9 | 4360 | 78 | ||
XM_007645221.1 | XP_007643411.1 | 8 - 9 | 4271 | 78 | ||
XM_007645222.1 | XP_007643412.1 | 7 - 8 | 4100 | 78 | ||
XM_007645223.1 | XP_007643413.1 | 7 - 8 | 2471 | 78 | ||
XM_007645224.1 | XP_007643414.1 | 8 - 9 | 2544 | 78 | ||
GenBank mRNA | JI880187.1 | - | 1 - 2 | 180 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |