Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511725.2 | XP_003511773.1 | 4 - 5 | 1206 | 74 | |
GenBank mRNA | JI893764.1 | - | 4 - 5 | 1203 | 74 | |
JP048637.1 | - | 4 - 5 | 1203 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |