Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511725.2 | XP_003511773.1 | 6 - 7 | 1369 | 95 | |
GenBank mRNA | JI893766.1 | - | 1 - 2 | 71 | 95 | |
JP048639.1 | - | 1 - 2 | 71 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |