Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511889.2 | XP_003511937.1 | 2 - 3 | 589 | 57 | |
XM_003511890.2 | XP_003511938.1 | 2 - 3 | 542 | 57 | ||
XM_007652381.1 | XP_007650571.1 | 3 - 4 | 734 | 57 | ||
GenBank mRNA | JI890996.1 | - | 2 - 3 | 430 | 57 | |
JP044362.1 | - | 2 - 3 | 430 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |