Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647072.1 | XP_007645262.1 | 3 - 4 | 669 | 71 | |
XM_007647073.1 | XP_007645263.1 | 5 - 6 | 483 | 71 | ||
XM_007647074.1 | XP_007645264.1 | 4 - 5 | 410 | 71 | ||
XM_007647075.1 | XP_007645265.1 | 4 - 5 | 429 | 71 | ||
XM_007647076.1 | XP_007645266.1 | 3 - 4 | 370 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |