Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hddc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652463.1 | XP_007650653.1 | 2 - 3 | 356 | 78 | |
GenBank mRNA | JP038704.1 | - | 2 - 3 | 356 | 78 |