Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500231.1 | XP_003500279.1 | 4 - 5 | 649 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |