Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507074.2 | XP_003507122.1 | 3 - 4 | 476 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |